Order Entry
Denmark
ContactUsLinkComponent
Dichloromethane-[D₂] 99.6+ atom % D for NMR spectroscopy
Irritant
Health
Dichloromethane-[D₂] 99.6+ atom % D for NMR spectroscopy
Catalog # ACRO217350100
CAS Number:  
undefined
Irritant
Health
Dichloromethane-[D₂] 99.6+ atom % D for NMR spectroscopy
Catalog # ACRO217350100
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    10 mL
  • Seq:
    2076
  • Pk Info:
    10 x 1 ml

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.04 % (Coulometric)
  • D-Enrichment:
    ≥99.60 %

Specification Test Results

About this item

 :
 :86.92 g/mol
Boiling Pt:40 °C (1013 hPa)
Melting Pt:–97 °C
Density:1.36 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:216-776-0
UN:1593
ADR:6.1,III
 :GHS07
 :GHS08
CAS Number:1665-00-5
Formula:CD₂Cl₂
MDL Number:MFCD00000882