Order Entry
Denmark
ContactUsLinkComponent
Dichloromethane-[D₂] 99.5 atom % D for NMR spectroscopy
Irritant
Health
Dichloromethane-[D₂] 99.5 atom % D for NMR spectroscopy
Certificates
undefined
Irritant
Health
Dichloromethane-[D₂] 99.5 atom % D for NMR spectroscopy
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.04 % (Coulometric)
  • D-Enrichment:
    ≥99.4 %
  • Identification C:
    Passes test
  • Identification D:
    Passes test
  • Alkalinity:
    Passes test
  • Titration:
    ≥85 %
  • NaCl and Na2SO4:
    ≤8 %
  • Non-esterified alcohols:
    ≤4 %