n-Pentane, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 8239.2500
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
- Environmentally Preferable:
Specifications
Specification Test Results
- Boiling Range:34 - 37 °C
Specification Test Results
About this item
Synonyms: Pentane
:
:72.15 g/mol
Merck Index:12,07255
Boiling Pt:36.1 °C (1013 hPa)
Melting Pt:–130 °C
Density:0.626 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-692-4
UN:1265
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:109-66-0
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498