

2,5-Dimethoxytetrahydrofuran for synthesis, Sigma-Aldrich®
: Merck
:696-59-3
:
:C₆H₁₂O₃
:MFCD00005359
:132.16 g/mol
:145 °C (1013 hPa)
:–45 °C
:1.021 g/cm³ (20 °C)
:35 °C
:1993
:3,III
:M-Clarity™ quality level = MQ200
:211-797-1
:GHS02
:GHS07
:Ambient
- Solubility:300 g/l (20 °C)
- Melting Point:-45 °C
- Molar Mass:132.15 g/mol
- Boiling Point:145 - 148 °C (1.013 hPa)
- Vapor Pressure:5 hPa (20 °C)
- Flash Point:35 °C
- Refractive Index:1.418 (20 °C, 589 nm)
- Explosion Limit:2.0 - 12.3 %(V)
- Density:1.02 g/cm3 (20 °C)
- pH:5 (300 g/l, H2O, 20 °C)
- Ignition Point:205 °C
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Frequently Bought Together