Order Entry
Denmark
ContactUsLinkComponent
2,5-Dimethoxytetrahydrofuran cis- and trans-mixture 99%
Toxic
Flammable
2,5-Dimethoxytetrahydrofuran cis- and trans-mixture 99%
Certificates
undefined
Toxic
Flammable
2,5-Dimethoxytetrahydrofuran cis- and trans-mixture 99%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4160 to 1.4190 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together