Order Entry
Denmark
ContactUsLinkComponent
2-Bromo-2-butene cis- and trans-mixture 98%
2-Bromo-2-butene cis- and trans-mixture 98%
Certificates
undefined
2-Bromo-2-butene cis- and trans-mixture 98%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear yellow to brown Liquid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4590 to 1.4620 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together