Rhenium ≥99.97% (metals basis), foil, annealed, Thickness: 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-15-5
:
Formula:Re
MDL Number:MFCD00011195
:186.21 g/mol
Merck Index:12,08343
Boiling Pt:5630 °C (1013 hPa)
Melting Pt:3180 °C
Density:21.02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-124-5
Frequently Bought Together