Rhenium ≥99.97% (metals basis), wire, annealed, Ø 0.1 mm (0.004 in)
Catalog # 10310.BY
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:100 cm
- Seq:7860
Specifications
About this item
:
:186.21 g/mol
Merck Index:12,08343
Boiling Pt:5630 °C (1013 hPa)
Melting Pt:3180 °C
Density:21.02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-124-5
CAS Number:7440-15-5
Formula:Re
MDL Number:MFCD00011195