Order Entry
Denmark
ContactUsLinkComponent
Rhenium ≥99.97% (metals basis), rods, Ø 4.7 - 5.0 mm
Rhenium ≥99.97% (metals basis), rods, Ø 4.7 - 5.0 mm
Catalog # 00270.BX
CAS Number:  
Rhenium ≥99.97% (metals basis), rods, Ø 4.7 - 5.0 mm
Catalog # 00270.BX
CAS Number:  

Specifications

  • Pk:
    12,5 mm
  • Seq:
    1625

Specifications

About this item

 :
 :186.21 g/mol
Merck Index:12,08343
Boiling Pt:5630 °C (1013 hPa)
Melting Pt:3180 °C
Density:21.02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-124-5
CAS Number:7440-15-5
Formula:Re
MDL Number:MFCD00011195