Order Entry
Denmark
Orders LinkContactUsLinkComponent
Kjeldahl catalyst, tablets

Kjeldahl catalyst, tablets

Kjeldahl Tablets ca. 1,0 g, Na2SO4; Se, General purpose, Aluminium tube, 1.000 Tab.
Catalog # 330666A
Supplier:   VWR Chemicals
Kjeldahl catalyst, tablets
Catalog # 330666A
Supplier:  VWR Chemicals
CAS Number:  
Kjeldahl catalyst, tablets
Kjeldahl catalyst, tablets

Kjeldahl Tablets ca. 1,0 g, Na2SO4; Se, General purpose, Aluminium tube, 1.000 Tab.

CAS Number:  
Price
4.570,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

This product is not available in all countries. Please check with your local Avantor International office or supplier.

Specifications

Specifications

  • Pack type:
    Aluminium tube
  • Pk:
    1.000 Tab.
  • Description:
    Kjeldahl Tablets ca. 1,0 g
  • For:
    General purpose
  • Contents:
    Na<sub>2</sub>SO<sub>4</sub>; Se

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.