Order Entry
Denmark
ContactUsLinkComponent
Cleanroom gloves, nitrile, sterile, Kimtech™ G3 Sterling™
Cleanroom gloves, nitrile, sterile, Kimtech™ G3 Sterling™
Supplier:  KIMBERLY CLARK
   Sustainable Options Available
Certificates
undefined
Cleanroom gloves, nitrile, sterile, Kimtech™ G3 Sterling™
Supplier:  KIMBERLY CLARK
   Sustainable Options Available
Certificates
Specifications
  • Catalog No:
  • 112-1864
  • 112-1865
  • 112-1862
  • 112-1863
  • 112-1861
  • 112-1858
  • 112-1859
  • 112-1860
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Cleanroom class:
  • 1/ISO 3
  • 1/ISO 3
  • 1/ISO 3
  • 1/ISO 3
  • 1/ISO 3
  • 1/ISO 3
  • 1/ISO 3
  • 1/ISO 3
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,07 mm
  • 0,07 mm
  • 0,07 mm
  • 0,07 mm
  • 0,07 mm
  • 0,07 mm
  • 0,07 mm
  • 0,07 mm
  • Finger thickness:
  • 0,10 mm
  • 0,10 mm
  • 0,10 mm
  • 0,10 mm
  • 0,10 mm
  • 0,10 mm
  • 0,10 mm
  • 0,10 mm
  • Palm thickness:
  • 0,08 mm
  • 0,08 mm
  • 0,08 mm
  • 0,08 mm
  • 0,08 mm
  • 0,08 mm
  • 0,08 mm
  • 0,08 mm
  • EN norm:
  • EN 374-1 Type C (K), EN 374-5 (VIRUS)
  • EN 374-1 Type C (K), EN 374-5 (VIRUS)
  • EN 374-1 Type C (K), EN 374-5 (VIRUS)
  • EN 374-1 Type C (K), EN 374-5 (VIRUS)
  • EN 374-1 Type C (K), EN 374-5 (VIRUS)
  • EN 374-1 Type C (K), EN 374-5 (VIRUS)
  • EN 374-1 Type C (K), EN 374-5 (VIRUS)
  • EN 374-1 Type C (K), EN 374-5 (VIRUS)
  • Anti-static:
  • +
  • +
  • +
  • +
  • +
  • +
  • +
  • +