Penicillinase (beta-Lactamase) (from Bacillus cereus)
Catalog # 426205-500
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pk:500 EU
- CAS No.:9001-74-5
- Source:Bacillus cereus
- Storage conditions:Refrigerator
- Enzyme name:Penicillinase (beta-Lactamase)