Ordreindtastning
Denmark
ContactUsLinkComponent
Methyl Violet 2B, ren, certified
undefined
Methyl Violet 2B, ren, certified
  • Appearance:
    Green Crystalline powder
  • Infrared spectrum:
    Conforms
  • Dye content:
    ≥75 %
  • UV Lambda max:
    582 to 588 nm
  • Absorption Ratio P-15/P+15:
    1.06 to 1.19
  • BSC Certification:
    Passes
Frequently Bought Together