Amberlite® IRN-78, Ion exchange resin cationic type (OH-), Supelco®
Catalog # 10343-U
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
CAS Number:11128-95-3
MDL Number:MFCD00145822