
Resazurin sodium salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:62758-13-8
:
Formula:C₁₂H₆NNaO₄
MDL Number:MFCD00005036
:251.17 g/mol
:M-Clarity™ quality level = MQ200
EINECS:263-718-5
:GHS07
Storage Temperature:Ambient
Frequently Bought Together