IR3535® (Insect repellent 3535)
Supplier: Merck
Certificates
About this item
Synonyms: IR3535® (Insect repellent 3535), 3-[N-n-butyl-N-acetyl] aminopropionic acid ethylester
CAS Number:52304-36-6
:
Formula:C₁₁H₂₁NO₃
MDL Number:MFCD00214076
Boiling Pt:292
Density:0.998 g/cm³ (20 °C)
Molecular Weight:215.29 g/mol
Flash Pt:159
Storage Temperature:Ambient
EINECS:257-835-0
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance :Clear, colorless to slightly yellowish liquid; almost odorless
- Identification (IR) :IR-Spectrum Conform;
- Assay :Not less than 99.0 %
- Appearance of sol (5%, water):Clear and colorless
- pH (5%, water) :4.0-6.0
- Free acid (as acetic acid):Not more than 0.1 %
- Water (K.F.) :Not more than 0.5 %
- Refractive index :1.452-1.455
- Sulphated ash :Not more than 0.1 %
- Total viable aerobic count:Not more than 100 CFU/g
- Escherichia coli:Absence in 1 g
- Pseudomonas aeruginos:Absence in 1 g
- Staphylococcus aureus :Absence in 1 g
- Candida albicans:Absence in 1 g
- Salmonella :Absence in 10 g
Specification Test Results
Related Information
Certificates
Search by Lot Numbermsds_111887
SDS
ProductPage_111887 spec_MERC1.11887.0000
Product specification
Frequently Bought Together