Docusate sodium
Glass bottle for solids, 500 g
Catalog # SERA20540.02
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate
- CAS Number:577-11-7
- Formula:C₂₀H₃₇NaO₇S
- MDL Number:MFCD00012455
- Molecular Weight:444.57 g/mol
- Merck Index:12,03460
- Boiling Pt:∼180 °C (1013 hPa)
- Melting Pt:∼155 °C
- Density:∼1.1 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:209-406-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



