Docusate sodium
Glass bottle for solids, 200 mg
Catalog # PROOBP133
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate
- CAS Number:577-11-7
- Formula:C₂₀H₃₇NaO₇S
- MDL Number:MFCD00012455
- Molecular Weight:444.57 g/mol
- Merck Index:12,03460
- Boiling Pt:∼180 °C (1013 hPa)
- Melting Pt:∼155 °C
- Density:∼1.1 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:209-406-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:200 mg



