Order Entry
Denmark
ContactUsLinkComponent
DIN adapter
DIN adapter
Catalog # 616-3177
Supplier:  AMETEK
undefined
DIN adapter
Catalog # 616-3177
Supplier:  AMETEK
Specifications
  • Description:
    DIN spindle adapter set
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Adapter for test geometry designed to provide an alternative for those customers with limited sample size. The cylindrical geometry provides defined shear rates.
  • Measuring range from 1 to 50000 mPa.s
  • Sample size typically of 16 to 20 ml
  • Easy to assemble and use
The sample can be held at moderate temperature using the jacketed chamber or alternatively can be immersed in a water bath.
Certifications: DIN 53019
Delivery: Supplied with three spindles (DIN 85, 86, 87) and corresponding chambers.