Order Entry
Denmark
ContactUsLinkComponent
Accessories for Transilluminators
Accessories for Transilluminators
  730-2902
Accessories for Transilluminators
  730-2902

 

  • Description:
    Medium wave tube, 302 nm, 8 W
  • Seq:
    02
  • Environmentally Preferable:
  • Pk:
    1