Order Entry
Denmark
Orders LinkContactUsLinkComponent
Plastic folding magnifiers
Plastic folding magnifiers
Supplier:   ESCHENBACH OPTIK
Plastic folding magnifiers
Supplier:  ESCHENBACH OPTIK
Certificates
Plastic folding magnifiers
Plastic folding magnifiers
2 Options Available Below
Product Details & Documents
These magnifiers featuring a modern design and a glass fibre reinforced PA body provide a distortion-free image.
  • For handicraft and technical applications
  • Dustproof body when closed
Ø: 23 mm
Product Family Options
Product Information
  • Magnification
    Ø
    Availability
    Price
  • 630-8260
    Achromatic double lens
    Achromatic double lens
    23 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.130,00 kr
    1.130,00 kr
    1 * 1 Items
       1.130,00 kr
  • 630-8261
    Aplanatic single lens
    Aplanatic single lens
    10×
    23 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 659,00 kr
    659,00 kr
    1 * 1 Items
       659,00 kr