Ice machine, cubes
Om denne vare
Ice machine in stainless steel for the production of ice cubes. Capacity from 18 to 145 kg/24 h. The ice machines are delivered with air cooling as a standard but are also available with water cooling. Water cooling is recommended for integrated machinery. Models with higher capacity are available. Please contact your local Avantor sales office for further details.
3 tilgængelige muligheder nedenfor
Product Details & Documents
Ice machine in stainless steel for the production of ice cubes. Capacity from 18 to 145 kg/24 h. The ice machines are delivered with air cooling as a standard but are also available with water cooling. Water cooling is recommended for integrated machinery. Models with higher capacity are available. Please contact your local Avantor sales office for further details.
Product Family Options
Produktinformation
- Ice capacity (l)Storage capacityRefrigerantW×D×HWeightAvailabilityPrice
- 474-0093Ice Machine for cubes, KL 22Ice Machine for cubes, KL 2224 - 25 kg/24 h6.5 kgR290345×460×588/613 mm32 kgLog ind for tilgængelighed1 * 1 Items11.670,00 kr
- 474-0094Ice Machine for cubes, KL 32Ice Machine for cubes, KL 3230 - 31 kg/24 h11 kgR290417×531×654/679mm39 kgLog ind for tilgængelighed1 * 1 Items14.480,00 kr
- 474-0095Ice Machine for cubes, KL 42Ice Machine for cubes, KL 4240 - 42 kg/24 h17 kgR290495×573×705/845mm49 kgLog ind for tilgængelighed1 * 1 Items17.030,00 kr