Ice machine, flakes
Ice flakers produce ice flakes that are dry and compact, this allows the ice to remain free flowing for easy use even after long storage periods. They have been manufactured to the highest specification, this includes a body work of high quality stainless steel grade 304 S 01. This eliminates any possibility of rusting when working in harsh environments. All internal components utilised are of either ABS (food grade) plastic or materials for use with potable water.
Used in many diverse areas: food, supermarket, laboratories, hopitals
- Air cooled
- Legs as standard
- Integral storage bins have stainless steel exterior and one piece ABS food safe plastic interiors for easy cleaning
The machines which are fully automatic are also fitted with mains on/off switches and a bin thermostat, which, on integral models prevents the system from producing ice when the storage bin is full. On modular machines the production of ice is controlled by an electronic eye, which detects when the storage bin is full.
With internal compartment in stainless steel for the production of flakes.
Height including legs.
Ice flakers produce ice flakes that are dry and compact, this allows the ice to remain free flowing for easy use even after long storage periods. They have been manufactured to the highest specification, this includes a body work of high quality stainless steel grade 304 S 01. This eliminates any possibility of rusting when working in harsh environments. All internal components utilised are of either ABS (food grade) plastic or materials for use with potable water.
Used in many diverse areas: food, supermarket, laboratories, hopitals
- Air cooled
- Legs as standard
- Integral storage bins have stainless steel exterior and one piece ABS food safe plastic interiors for easy cleaning
The machines which are fully automatic are also fitted with mains on/off switches and a bin thermostat, which, on integral models prevents the system from producing ice when the storage bin is full. On modular machines the production of ice is controlled by an electronic eye, which detects when the storage bin is full.
With internal compartment in stainless steel for the production of flakes.
Height including legs.
Product Family Options
- DescriptionIce capacity (l)Storage capacityRefrigerantW×D×HWeight
- Automatic ice flake machine, KF 4540 - 42 L/24 h10 kgR290345×520×610/635 mm37 kg1 * 1 Items23.420,00 kr
- Automatic ice flake machine, KF 8580 - 83 L/24 h25 kgR290485×586×720/860 mm54 kg1 * 1 Items27.080,00 kr
- Automatic ice flake machine, KF 145135 - 141 L/24 h42 kgR290680×588×890/1030 mm69 kg1 * 1 Items28.870,00 kr
- Automatic ice flake machine, KF 185175 - 183 L/24 h60 kgR290953×590×781/921 mm81 kg1 * 1 Items37.890,00 kr
- Automatic Ice flake machine, KF7575 - 78 L/24 h75 kg485×585×720 mm54 kg-