Order Entry
Denmark
Orders LinkContactUsLinkComponent
Hotplates
Hotplates
Supplier:   NICKEL ELECTRO
Hotplates
Supplier:  NICKEL ELECTRO
Certificates
Hotplates
3 Options Available Below
Product Details & Documents
Stainless steel casing, stove enamelled finish and cast aluminium plate. Units accept retort rods.
  • Temperature controlled by hydraulic thermostat
  • With illuminated on/off switch and heater “ON” indicator
  • Fitted with non-slip rubber feet for safety
Each hotplate accepts an optional sand bath attachment with aluminium upstand rim which fits over edge of hotplate. The attachment can also act as a safety rim to prevent dishes falling off hotplate.
Product Family Options
Product Information
  • Heating area
    Temp.
    Heating capacity
    Plate size
    Type
    W×D×H
    Availability
    Price
  • 305×240 mm
    Max. 350 °C
    1000 W
    305×240 mm
    HP1-1
    305×240×200 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 12.530,00 kr
    12.530,00 kr
    1 * 1 Items
       12.530,00 kr
  • 457×305 mm
    Max. 350 °C
    2000 W
    457×305 mm
    HP1-2
    457×305×200 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 15.480,00 kr
    15.480,00 kr
    1 * 1 Items
       15.480,00 kr
  • 686×305 mm
    Max. 350 °C
    3000 W
    686×305 mm
    HP1-3
    690×315×200 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 19.600,00 kr
    19.600,00 kr
    1 * 1 Items
       19.600,00 kr