Order Entry
Denmark
ContactUsLinkComponent
 
Bottle-top dispenser, ceramus® HF
  613-3238
undefined
Bottle-top dispenser, ceramus® HF
  613-3238

 

  • Volume:
    2 - 10 ml
  • Accuracy:
    ±0,6 %
  • Division:
    0,25 ml
  • Imprecision:
    <0,2 %
  • Pk:
    1

 

 

Variable volume, autoclavable.

  • Multipurpose dispenser for hydrofluoric acid and very aggressive liquids
  • Micrometric adjustment knob for fine tuning
  • Piston, cylinder, seat and valve ball made from pure ceramic, valve spring made from platinum/iridium
  • Convenient recirculation system for rapid flushing with no loss of reagents
  • Certification of quality and conformity
 : Supplied with A 45 thread, three adapters (A 32, A 38, S 40), one aspiration tube with screw connection, serial number and quality certificate.