WHEATON® Serum Vials
Supplier: DWK Life Sciences
Certificates
About this item
These clear glass vials are manufactured from Wheaton '200' low extractable borosilicate glass to maintain uniform wall thickness. The amber vials are manufactured from Wheaton '320' borosilicate glass.
- Blowback feature in the neck and a specially designed bottom radius for added strength
- These features are important for freeze-drying applications
- Vials are provided in shrink-wrapped modules which greatly reduces particulate contamination
Certifications: Meets ASTM Type 1, Class A/B and USP Type 1 standards.
Delivery information: Vials are supplied without closures.