Test tubes with standard ground stoppers
Leverandør: SciLabware
Gå til:
Om denne vare
Pyrex® borosilicate clear glass.
- Graduated
- Excellent chemical and thermal resistance
- Interchangeable PE stopper (included)
Ground socket tubes with white enamel graduations to indicate capacity.
Certifications: ISO 4142, DIN 12217
4 tilgængelige muligheder nedenfor
Product Details & Documents
Pyrex® borosilicate clear glass.
- Graduated
- Excellent chemical and thermal resistance
- Interchangeable PE stopper (included)
Ground socket tubes with white enamel graduations to indicate capacity.
Certifications: ISO 4142, DIN 12217
Product Family Options
Produktinformation
- CapacityLengthExt.ØSocket sizeDivisionTilgængelighedPrice
Anbefalinger