Microcentrifuge tubes, 2,0 ml
Katalog # 211-2606
Supplier: Brand
Specifications
- Type:with attached lid
- Version:graduated, conical bottom, non sterile
- Capacity:2,0 ml
- Colour:Colourless
- Pk:500
Specifications
About this item
PP, graduated, autoclavable with attached cap.
- Can withstand centrifugation up to 20000×g
- Flat cap can be easily labelled and pierced
- Frosted marking area