Culture tubes with screw cap
Supplier: Brand
Certificates
About this item
AR® glass, PP screw cap and white elastomer seal.
- Autoclavable (+121 °C)
- Wall thickness 1 mm
- GL thread
Frequently Bought Together
AR® glass, PP screw cap and white elastomer seal.