VWR®, Centrifuge Tubes, High Performance
Supplier: VWR Collection
Certificates
About this item
PP, conical, with purple HDPE flat cap and integral sealing or plug-style cap and deep sealing area.
- Certified non cytotoxic, nuclease- and pyrogen-free
- Caps available in two styles: flat cap with integral sealing and a smooth surface for additional labelling options, or plug-style cap with deep sealing area for a secure seal when using shakers and horizontal incubators
- Easy to read black graduations and white writing area on the side for permanent coding
- Can be used for storing and freezing samples at temperatures down to –80 °C
- Can be centrifuged up to 12500×g
- Sustainable design based on virgin resins
These disposable, conical-bottom, free-standing tubes are extremely versatile, and can be used for routine tasks as well as high speed centrifuging. Rack-packed tubes are packed in freezable, recyclable, autoclavable racks. Tubes are sterilised by gamma irradiation (except free-standing tubes in bags or bulk). Tubes and caps are autoclavable and freezable.
Packaging: Bulk, in bags, or in autoclavable plastic racks.
Specifications
- Catalog No:
- 525-0402
- 525-0400
- Environmentally Preferable:
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
Specifications
Related Information
Certificates
Search by Lot NumberCentrifugation Solutions from Avantor VWR for Cell Culture
Product Literature
Frequently Bought Together