Centrifuge tubes, conical
Supplier: DELTALAB
Jump to:
About this item
Tubes made of PP, translucent, with blue HDPE screw cap.
- Continuous thread
- External moulded graduations in 0,5 ml increments
- Excellent temperature resistance
- Excellent chemical resistance
Suitable for centrifugation tests in immunology, microbiology, more.
9 Options Available Below
Product Details & Documents
Tubes made of PP, translucent, with blue HDPE screw cap.
- Continuous thread
- External moulded graduations in 0,5 ml increments
- Excellent temperature resistance
- Excellent chemical resistance
Suitable for centrifugation tests in immunology, microbiology, more.
Product Family Options
Centrifuge tubes 15 ml
Product Information
- CapacityØ×LSterileClosure typeRCF max.TypeVersionPackedAvailabilityPrice
- 525-4214Centrifuge tubes, non-sterileCentrifuge tubes, non-sterile15 ml17×122 mmScrew cap7000×gWithout skirtNon sterileBulkLog in for availability1 * 500 Items884,00 kr
- 15 ml17×122 mmScrew cap5000×gWithout skirtSterile100 per bag/500 per caseLog in for availability1 * 500 Items1.460,00 kr
- 15 ml17×122 mmScrew cap5000×gWithout skirtSterileIndividually baggedLog in for availability1 * 500 Items1.450,00 kr
Centrifuge tubes 50 ml
Product Information
- CapacityØ×LSterileClosure typeRCF max.TypeVersionPackedAvailabilityPrice
- 50 ml29×118,5 mmScrew cap12000×gWithout skirtNon sterileBulkLog in for availability1 * 500 Items1.150,00 kr
- 525-4217Centrifuge tubes, skirted, non-sterileCentrifuge tubes, skirted, non-sterile50 ml29×118,5 mm-12000×gSkirtedNon sterileBulkLog in for availability1 * 500 Items1.230,00 kr
- 50 ml29×118,5 mmScrew cap7000×gWithout skirtSterileIndividually baggedLog in for availability1 * 500 Items1.970,00 kr
- 525-4219Centrifuge tubes, skirted, sterileCentrifuge tubes, skirted, sterile50 ml29×118,5 mm+Screw cap7000×gSkirtedSterileIndividually baggedLog in for availability1 * 500 Items2.190,00 kr
- 50 ml29×118,5 mmScrew cap7000×gSkirtedSterile100 per bag/500 per caseLog in for availability1 * 500 Items2.160,00 kr
- 50 ml29×118,5 mmScrew cap7000×gSkirtedSterile25 per bag/500 per caseLog in for availability1 * 500 Items2.250,00 kr