Order Entry
Denmark
ContactUsLinkComponent
Sterilising pan, Nalgene®
Sterilising pan, Nalgene®
Catalog # 489-2110
   Low Manufacturing Impact
undefined
Sterilising pan, Nalgene®
Catalog # 489-2110
   Low Manufacturing Impact
Specifications
  • W×D×H:
    152×456×67 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Pk:
    1
Specifications
About this item
PP with cover.
  • Cover minimises spillage of hazardous materials
  • Resistant to aggressive disinfectants
  • Can also be used to sterilise labware instruments and utensils
  • Stackable
Allows pipettes to be fully immersed in disinfectant.
Meets OSHA Standard 29 CFR (Part 1910.1030) for use as protection against blood-borne pathogens.
Certifications: Meets OSHA Standard 29 CFR (Part 1910.1030) for use as protection against blood-borne pathogens.