Order Entry
Denmark
ContactUsLinkComponent
Labelling tape cartridges, TZ, for label printer P-Touch
Labelling tape cartridges, TZ, for label printer P-Touch
Catalog # 811-0873
Supplier:  BROTHER
undefined
Labelling tape cartridges, TZ, for label printer P-Touch
Catalog # 811-0873
Supplier:  BROTHER

Specifications

  • Printer model:
  • Printing method:
  • Colour:
    White/blue
  • Description:
    Labelling PET tape cartridge, TZ tape for P-Touch
  • Dimensions:
    18 mm×8 m
  • Material:
    PET
  • Environmentally Preferable:
  • Pk:
    1 Roll

Specifications

About this item

These TZ labelling tape are developped for use with P-Touch labelprinters.

  • Normal laminated tape
  • Many different colours

Length: 8 m