Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cleanroom tapes, Supertack™
Cleanroom tapes, Supertack™
Supplier:   ULTRATAPE
Cleanroom tapes, Supertack™
Supplier:  ULTRATAPE
Certificates
Cleanroom tapes, Supertack™
Cleanroom tapes, Supertack™
17 Options Available Below
Product Details & Documents
These Supertack™ cleanroom tapes are made of PE with rubber adhesive and are ideal for any application that requires a strong bond.
  • Tapes remove cleanly with no residue
  • Tear by hand
  • Accept cleanroom ballpoint pen writing
The tape is available in Protocol™ uniquely striped color pattern which allows immediate recognition of the product as a certified cleanroom tape. Suitable for use in ISO Class 4 environments
Length: 32,9 m.
Different colours available.
Packaging: Supplied double-bagged.
Documents
Product Family Options
Product Information
  • Colour
    Width
    Availability
    Price
  • UTAP0153BL050-P3S
    Cleanroom tape
    Cleanroom tape
    Blue
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 48 ROL
    1 * 48 ROL - 6.520,00 kr
    6.520,00 kr
    1 * 48 ROL
       6.520,00 kr
  • UTAP0153BW100-P3S
    Cleanroom tape
    Cleanroom tape
    Blue/white
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 13 ROL
    1 * 13 ROL - 2.170,00 kr
    2.170,00 kr
    1 * 13 ROL
       2.170,00 kr
  • UTAP1153BW100-P3D
    Cleanroom tape
    Cleanroom tape
    Blue/white
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 12 ROL
    1 * 12 ROL - 2.390,00 kr
    2.390,00 kr
    1 * 12 ROL
       2.390,00 kr
  • UTAP1153WH100-P3DU
    Cleanroom tape
    Cleanroom tape
    White
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 6 ROL
    1 * 6 ROL
  • UTAP1153YE100P3D
    Cleanroom tape
    Cleanroom tape
    Yellow
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 12 ROL
    1 * 12 ROL - 2.390,00 kr
    2.390,00 kr
    1 * 12 ROL
       2.390,00 kr
  • UTAP0153BL200-P3S
    Cleanroom tape
    Cleanroom tape
    Blue
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 24 ROL
    1 * 24 ROL - 5.630,00 kr
    5.630,00 kr
    1 * 24 ROL
       5.630,00 kr
  • UTAP0153BW200-P3S
    Cleanroom tape
    Cleanroom tape
    Blue/white
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 9 ROL
    1 * 9 ROL - 2.110,00 kr
    2.110,00 kr
    1 * 9 ROL
       2.110,00 kr
  • UTAP1153BW200-P3D
    Cleanroom tape, Protocol™
    Cleanroom tape, Protocol™
    Blue/white
    51,8 mm
    Log in for availability
    Log in for availability
    1 * 9 ROL
    1 * 9 ROL - 3.030,00 kr
    3.030,00 kr
    1 * 9 ROL
       3.030,00 kr
  • UTAP1153YE200-P3D
    Cleanroom tape
    Cleanroom tape
    Yellow
    51,8 mm
    Log in for availability
    Log in for availability
    1 * 3 ROL
    1 * 3 ROL - 1.010,00 kr
    1.010,00 kr
    1 * 3 ROL
       1.010,00 kr
  • UTAP1153WH300-P3D
    Cleanroom tape
    Cleanroom tape
    White
    76,2 mm
    Log in for availability
    Log in for availability
    1 * 6 ROL
    1 * 6 ROL - 2.580,00 kr
    2.580,00 kr
    1 * 6 ROL
       2.580,00 kr
  • UTAP1153BL600-P3D
    Cleanroom tape
    Cleanroom tape
    Blue
    152,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 757,00 kr
    757,00 kr
    1 * 1 ROL
       757,00 kr
  • UTAP1153BW600-P3D
    Cleanroom tape, Protocol™
    Cleanroom tape, Protocol™
    Blue/white
    152,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 757,00 kr
    757,00 kr
    1 * 1 ROL
       757,00 kr
  • UTAP1153GR600-P3D
    Cleanroom tape
    Cleanroom tape
    Green
    152,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 757,00 kr
    757,00 kr
    1 * 1 ROL
       757,00 kr
  • UTAP1153OR600-P3D
    Cleanroom tape
    Cleanroom tape
    Orange
    152,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 757,00 kr
    757,00 kr
    1 * 1 ROL
       757,00 kr
  • UTAP1153RE600-P3D
    Cleanroom tape
    Cleanroom tape
    Red
    152,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 757,00 kr
    757,00 kr
    1 * 1 ROL
       757,00 kr
  • UTAP1153WH600-P3D
    Cleanroom tape
    Cleanroom tape
    White
    152,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 757,00 kr
    757,00 kr
    1 * 1 ROL
       757,00 kr
  • UTAP1153YE600-P3D
    Cleanroom tape
    Cleanroom tape
    Yellow
    152,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 757,00 kr
    757,00 kr
    1 * 1 ROL
       757,00 kr