Order Entry
Denmark
ContactUsLinkComponent
Swabs, Steriswabs®
Swabs, Steriswabs®
Catalog # 720-0142
undefined
Swabs, Steriswabs®
Catalog # 720-0142
Specifications
  • Description:
    Swabs in labelled tubes
  • Material:
    Plastic
  • Cap colour:
    Blue
  • Pk:
    250
Specifications
About this item
Swabs with 45 mm breakpoint, dry or pre-moistened with sterile diluent.
  • Available in labelled tube or peel pouches
  • Triple wrapped and radiation sterilised
Designed for effective monitoring of sterility and contamination in clinically clean or sterile work areas.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904