Beaker and flask holders, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Lead ring, with red vinyl coating and four stainless steel brackets. Holds beakers and flasks firmly in a fixed position. Ideal for use in water baths as it allows liquid to circulate beneath the vessel base.
- Good chemical resistance
- Good thermal resistance
- Easy, simple handling
Frequently Bought Together