Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sterilisation indicators

Sterilisation indicators

Tape, 50 m, 19 mm
  489-0046
Sterilisation indicators
  489-0046
Sterilisation indicators
Sterilisation indicators

Tape, 50 m, 19 mm

 
493,00 kr
   
1 * 1 ROL
 

Tapes indicators by dry heat sterilisation.

The tape (ethylene oxide) changes from white to black after 3,5 minutes at 134 °C.

Dry heat steriliser tape (poupinel) changes from green to different shades of brown after a period at a temperature between 155 and 180 °C.

 
  • Width:
    19 mm
  • Description:
    Tape
  • Length:
    50 m
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.