Order Entry
Denmark
Orders LinkContactUsLinkComponent
Spoon spatulas
Spoon spatulas
Supplier:   BOCHEM
Spoon spatulas
Supplier:  BOCHEM
Certificates
Spoon spatulas
6 Options Available Below
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic. Multipurpose spoon for a wide range of uses.
  • Round handle
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
Product Family Options
Product Information
  • Length
    Blade length
    Blade width
    Spoon size
    Availability
    Price
  • 120 mm
    35 mm
    10 mm
    30×22 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 49,50 kr
    49,50 kr
    1 * 1 Items
       49,50 kr
  • 150 mm
    40 mm
    10 mm
    30×22 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 52,40 kr
    52,40 kr
    1 * 1 Items
       52,40 kr
  • 180 mm
    50 mm
    12 mm
    40×29 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 55,40 kr
    55,40 kr
    1 * 1 Items
       55,40 kr
  • 210 mm
    50 mm
    12 mm
    40×29 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 62,10 kr
    62,10 kr
    1 * 1 Items
       62,10 kr
  • 250 mm
    70 mm
    14 mm
    48×35 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 66,00 kr
    66,00 kr
    1 * 1 Items
       66,00 kr
  • 300 mm
    70 mm
    14 mm
    48×35 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 70,00 kr
    70,00 kr
    1 * 1 Items
       70,00 kr