Order Entry
Denmark
Orders LinkContactUsLinkComponent
Holders for test tubes
Holders for test tubes
Supplier:   KARTELL
Holders for test tubes
Supplier:  KARTELL
Certificates
Holders for test tubes
3 Options Available Below
Product Details & Documents
PP, white.
  • Good thermal and chemical resistance
  • Can be used in water baths
  • The tubes contents are always visible
Designed for titrations.
Product Family Options
Product Information
  • No. of Holes
    For tube Ø
    W×D×H
    Availability
    Price
  • 10
    16 mm
    220×55×62,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 113,00 kr
    113,00 kr
    1 * 1 Items
       113,00 kr
  • 9
    18 mm
    220×55×65,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 124,00 kr
    124,00 kr
    1 * 1 Items
       124,00 kr
  • 8
    20 mm
    220×54×65,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 131,00 kr
    131,00 kr
    1 * 1 Items
       131,00 kr
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Base plate that can hold two racks, 202×156×13,5 mm
    Base plate that can hold two racks, 202×156×13,5 mm
    212-6897
    Log in for availability
    1 * 1 Items - 90,30 kr