TLC silica gel 60, unmodified layers, glass backed, LuxPlate®
Supplier: Merck
Certificates
About this item
The LuxPlate® range of high fluorescence TLC plates contains an increased amount of fluorescent dye making it easier to identify small amounts of isolated material. Furthermore, the amount of binder added is elevated, resulting in a significantly brighter TLC plate with an even more stable and robust surface.
- Twice as bright under UV light compared to conventional TLC plates
- Easy identification of the developed bands due to better contrast against plate background
Frequently Bought Together