Order Entry
Denmark
Orders LinkContactUsLinkComponent
Pipette tips, Combitips® for Multipette® 4780
Pipette tips, Combitips® for Multipette® 4780
12,5 ml, -, E, MP, MD, Bag
Catalog # 613-3512
Supplier:   EPPENDORF
Pipette tips, Combitips® for Multipette® 4780
Catalog # 613-3512
Supplier:  EPPENDORF
Pipette tips, Combitips® for Multipette® 4780
Pipette tips, Combitips® for Multipette® 4780
12,5 ml, -, E, MP, MD, Bag
Price
Inquire For Price
Log in for availability
1 * 100 Items
Product Details & Documents
Accurate, reliable dispensing system used in conjunction with the old Multipette® 4780.
Ordering information: 0,05 ml Combitips® are supplied in sets of 50 with 96×10 µl pipette tips.
Documents
Specifications
  • Packed:
    Bag
  • Sterile:
    -
  • Volume:
    12,5 ml
  • For use with:
    E, MP, MD