Special pipette tips, Finntip™
High quality PP, autoclavable.
- Finntip™ extended (200 and 1000 µl): long, tapered tip especially adapted for deep tubes and narrow necked bottles
- Finntip™ wide: tips with a wide, ground orifice for pipetting viscous liquids and fragile suspensions of cells or macromolecules, such as genomic DNA, without fragmentation
- Catalog No:
- 613-2652
- 613-2650
- 613-2648
- 613-4489
- 613-4488
- 613-2653
- 613-2620
- 613-4486
- 613-4487
- 613-2593
- 613-2592
- 613-2591
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
Frequently Bought Together