Infectious products shippers
Supplier: E3 Cortex
Certificates
About this item
Micro-fluted cardboard treated against moisture (Biotainer); outer double fluted carton (Cryopack®).
- Cardboard wedge
- Sheet of absorbent cloth
- Watertight secondary PP container
These packages are designed for the transport of infected or contaminated consumables. They are suitable for applications in room temperature (Biotainer®). They can also be used in controlled temperature of 1 to 8 °C or in freezing temperatures with dry ice (Cryopack®).
Approved by the UN. Comply with international regulations in force IATA class 6.2 IMDG and ADR RIG.
Certifications: Approved by the UN. Comply with international regulations in force IATA class 6.2 IMDG and ADR RIG.
Frequently Bought Together