Mops, MegaSwat™
Supplier: MICRONOVA
Certificates
About this item
These flat, paddle-shaped mops are for sliding behind heavy cabinets and process equipment, over piping and underneath calibrated equipment. Mop heads measure (W×D) 406×3 mm.
- Flexible polyethylene interior, choice of PVA, NovaLite latex rayon or NovaPoly polyester covering
- MegaSwat™ Lite with NovaLite cover withstands stronger disinfectants and can be autoclaved
- Mop heads attach to universal stainless steel handle via quick disconnect MegaSwat™ adapter
Ordering information: All components must be ordered separately.
Specifications
- Catalog No:
- MICRMS6-18NSIR
- MICRMSAA-16NS
- 129-1083
- MICRMS6-8NSIR
- 129-0518
- MICRMS6-18NS
- 129-0614
- MICRMS1111-16
- MICRMS6-16NSIR
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing