Deep well plates and cover mats
: Brand
PP or PS, non sterile. Designed for a wide range of applications, such as high throughput screening, cell and tissue cultures, serial dilutions, reagent transfer, and sample storage down to −80 °C (PP) or −20 °C (PS). Deep well plates are manufactured in standard ANSI/SLAS 1 and 4 format and are stackable for easy storage.
- PP for high chemical resistance, for example, against DMSO, phenol, chloroform
- Autoclavable at 121 °C (2 bar) acc. DIN EN 285 (PP plates only)
- Round shape wells allow optimal sample mixing and recovery
- Alphanumeric coding and cut-away corner simplify sample identification and orientation
- Can be used with multi channel pipettes and automatic liquid handling systems from all leading manufacturers
- Free from DNA, DNase and RNase (except low profile plate which is not tested)
Frequently Bought Together