Microplates, 1536-well
Supplier: Greiner Bio-One
About this item
PS, without lid. Ideal for high throughput screening applications, these plates are available as clear bottom variants (in clear PS) and black or white in LUMITRAC™ or FLUOTRAC™ quality.
- HiBase versions designed for top reading instruments, µClear® LoBase plates for bottom reading instruments
- Solid bottom plates available with high binding (MICROLON™ 600, LUMITRAC™ 600 or FLUOTRAC™ 600) or medium binding (LUMITRAC™ 200 or FLUOTRAC™ 200) surfaces
- Rounded square well design combines the advantages of the square well (flexible working volume, 3 to 10 µl) with the advantages of a round well (reduced wicking and bubbling)
- Free of DNase, RNase and human DNA, and are non-pyrogenic
Working volume: 3 - 10 µl
10 Options Available Below
Product Details & Documents
PS, without lid. Ideal for high throughput screening applications, these plates are available as clear bottom variants (in clear PS) and black or white in LUMITRAC™ or FLUOTRAC™ quality.
- HiBase versions designed for top reading instruments, µClear® LoBase plates for bottom reading instruments
- Solid bottom plates available with high binding (MICROLON™ 600, LUMITRAC™ 600 or FLUOTRAC™ 600) or medium binding (LUMITRAC™ 200 or FLUOTRAC™ 200) surfaces
- Rounded square well design combines the advantages of the square well (flexible working volume, 3 to 10 µl) with the advantages of a round well (reduced wicking and bubbling)
- Free of DNase, RNase and human DNA, and are non-pyrogenic
Working volume: 3 - 10 µl
Specifications for Product Family
Specifications
- Catalog No:
- 392-0138
- 738-0067
- 738-0070
- 738-0068
- 738-0069
- 738-0077
- 738-0078
- 738-0080
- 738-0089
- 738-0084
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
Specifications
Product Family Options
1536-well microplates, HiBase
Product Information
- ColourNo. of wellsSterileAvailabilityPrice
- 392-0138Solid bottom, clear, MICROLON 600, sterileSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.392-0138Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 738-0067Solid bottom, white, LUMITRAC 600, sterileSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.738-0067Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 738-0070Solid bottom, black, FLUOTRAC 600, sterileSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.738-0070Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 738-0068Solid bottom, white, LUMITRAC 200Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.738-0068Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 738-0069Solid bottom, black, FLUOTRAC 200Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.738-0069Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 738-0077µClear bottom, white, medium bindingSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.738-0077Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 738-0078µClear bottom, black, medium bindingSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.738-0078Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 738-0080Solid bottom, clearSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.738-0080Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact
1536-well microplates, LoBase
Product Information
- ColourNo. of wellsSterileAvailabilityPrice
- 738-0089µClear bottom, black, medium bindingSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.738-0089Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 738-0084Solid bottom, black, FLUOTRAC 200Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.738-0084Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact