Measuring cylinders, low form, class B, borosilicate glass
Leverandør: HIRSCHMANN
Gå til:
Om denne vare
DURAN®, borosilicate glass, low form, class B.
- Calibrated to contain 'In'
- Hexagonal base with spout
- Marks and inscriptions in amber stain, short graduation marks
DIN EN ISO 4788
8 tilgængelige muligheder nedenfor
Product Details & Documents
DURAN®, borosilicate glass, low form, class B.
- Calibrated to contain 'In'
- Hexagonal base with spout
- Marks and inscriptions in amber stain, short graduation marks
DIN EN ISO 4788
Documents
HIRS2400190_TDS_EN
Datasheet
Product Family Options
Produktinformation
- CapacityDivisionHeightToleranceTilgængelighedPrice