Reusable half masks, 6000 and 7000
Supplier: 3M
Certificates
About this item
Reusable, lightweight and comfortable to wear. Incorporating a soft, hypoallergenic face piece material and central exhalation valve.
- Uses 6000/2000 series bayonet filters
- Low maintenance respirator can be disassembled, cleaned and reused many times
- No replacement parts are necessary except filters and cartridges
EN 140
Certifications: EN 140
Ordering information: Supplied without filters, which are sold separately (except 111-0465, 111-0466).