Order Entry
Denmark
Orders LinkContactUsLinkComponent
Staining jars acc. to Coplin
Staining jar according to Coplin
  631-9331
Staining jars acc. to Coplin
  631-9331
Staining jars acc. to Coplin
Staining jars acc. to Coplin
Staining jar according to Coplin
249,00 kr
1 * 1 Items
Soda-lime glass.
For 10 standard microscopy slides (76×26 mm).
  • Description:
    Staining jar according to Coplin