Order Entry
Denmark
Orders LinkContactUsLinkComponent
Disposable base moulds
Disposable base moulds
Supplier:   Simport Scientific
Disposable base moulds
Supplier:  Simport Scientific
Certifikater
Disposable base moulds
Disposable base moulds
6 tilgængelige muligheder nedenfor
Product Details & Documents
These disposable PVC base moulds offer ease and convenience of operation. They can be discarded after use, but are also strong enough to be reused.
  • Excellent thermal exchange
  • Smooth interior
  • Rounded corners
They are available in the same variety of sizes as metal moulds and can be used with the same styles or types of cassettes and embedding rings.
Delivery information: Each case contains two dispenser boxes of 500 base moulds.
Product Family Options
Produktinformation
  • Dimensions
    Availability
    Price
  • 720-0820
    Base mould
    Base mould
    7×7×5 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 2.330,00 kr
    2.330,00 kr
    1 * 1.000 Items
       2.330,00 kr
  • 720-0821
    Base mould
    Base mould
    15×15×5 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.700,00 kr
    3.700,00 kr
    1 * 1.000 Items
       3.700,00 kr
  • 720-0822
    Base mould
    Base mould
    24×24×5 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.680,00 kr
    3.680,00 kr
    1 * 1.000 Items
       3.680,00 kr
  • 720-0823
    Base mould
    Base mould
    30×24×5 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 2.330,00 kr
    2.330,00 kr
    1 * 1.000 Items
       2.330,00 kr
  • 720-0824
    Base mould
    Base mould
    37×24×5 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 2.330,00 kr
    2.330,00 kr
    1 * 1.000 Items
       2.330,00 kr
  • 720-0208
    Deep base mould, for M512 MACROSETTE®
    Deep base mould, for M512 MACROSETTE®
    37×24×10 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 989,00 kr
    989,00 kr
    1 * 500 Items
       989,00 kr